1-(2-Methoxyethyl)piperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20286G25 AS20286G5
ACTIAS20286G25EA 0 GBP
ACTIAS20286G25 ACTIAS20286G5
1-(2-Methoxyethyl)piperazine
1-(2-Methoxyethyl)piperazine
Formula: C₇H₁₆N₂O
MDL Number: MFCD00191214
CAS Number: 13484-40-7
UN: 3267
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR