2'-Hydroxy-4'-methoxyacetophenone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24060G100
AS24060G25
ACTIAS24060G100EA
0
GBP
ACTIAS24060G100
ACTIAS24060G25
2'-Hydroxy-4'-methoxyacetophenone
2'-Hydroxy-4'-methoxyacetophenone
Formula:
C₉H₁₀O₃ Boiling Pt: 154 °C (20 mmHg) Melting Pt: 51 °C Density: 1,1604 g/cm³ (40 °C) |
MDL Number:
MFCD00008730 CAS Number: 552-41-0 EINECS: 209-012-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...