ω-Bromo-2-nitroacetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: α-Bromo-2-nitroacetophenone, 2-Bromo-2'-nitroacetophenone, Bromomethyl o-nitrophenyl ketone, 2-Nitrophenacyl bromide

AS20334G100 AS20334G25 AS20334G5
ACTIAS20334G100EA 0 GBP
ACTIAS20334G100 ACTIAS20334G25 ACTIAS20334G5
ω-Bromo-2-nitroacetophenone
ω-Bromo-2-nitroacetophenone
Formula: C₈H₆BrNO₃
MDL Number: MFCD00010294
CAS Number: 6851-99-6
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR