2-Methoxy-4-nitrophenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21371G100 AS21371G25
ACTIAS21371G100EA 0 GBP
ACTIAS21371G100 ACTIAS21371G25
2-Methoxy-4-nitrophenol
2-Methoxy-4-nitrophenol
Formula: C₇H₇NO₄
Melting Pt: 103…104 °C
MDL Number: MFCD00012143
CAS Number: 3251-56-7
EINECS: 221-839-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR