4-Bromo-2-methoxyphenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromoguaiacol, 5-Brom-2-hydroxyanisole

AS21334G25 AS21334G5
ACTIAS21334G25EA 57 GBP
ACTIAS21334G25 ACTIAS21334G5
4-Bromo-2-methoxyphenol
4-Bromo-2-methoxyphenol
Formula: C₇H₇BrO₂
MW: 203.04 g/mol
MDL Number: MFCD00051937
CAS Number: 7368-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR