2-Bromo-4-nitrophenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Bromo-2-hydroxy-5-nitrobenzene

AS22589G1 AS22589G25 AS22589G5
ACTIAS22589G1EA 0 GBP
ACTIAS22589G1 ACTIAS22589G25 ACTIAS22589G5
2-Bromo-4-nitrophenol
2-Bromo-4-nitrophenol
Formula: C₆H₄BrNO₃
MW: 218.01 g/mol
MDL Number: MFCD06656567
CAS Number: 5847-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR