tert-Butyl carbamate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Carbamic acid tert-butyl ester
AS23105G25
AS23105G100
AS23105G5
ACTIAS23105G25EA
22
GBP
ACTIAS23105G25
ACTIAS23105G100
ACTIAS23105G5
tert-Butyl carbamate
tert-Butyl carbamate
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol |
MDL Number:
MFCD00007962 CAS Number: 4248-19-5 EINECS: 224-209-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...