(R)-(-)-3-Chloro-1,2-propanediol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (R)-(-)-3-Chloro-1,2-dihydroxy propane, (2R)-(-)-3-Chloropropane-1,2-diol

AS20183G100 AS20183G25
ACTIAS20183G100EA 0 GBP
ACTIAS20183G100 ACTIAS20183G25
(R)-(-)-3-Chloro-1,2-propanediol
(R)-(-)-3-Chloro-1,2-propanediol
Formula: ClCH₂CHOHCH₂OH
MW: 110.54 g/mol
Boiling Pt: 74…76 °C (15 mmHg)
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00135169
CAS Number: 57090-45-6
EINECS: 435-560-9
UN: 2689
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR