2-Chloro-3-methylthiophene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS25242G100 AS25242G25
ACTIAS25242G100EA 212 GBP
ACTIAS25242G100 ACTIAS25242G25
2-Chloro-3-methylthiophene
2-Chloro-3-methylthiophene
Formula: C₅H₅ClS
MW: 132.61 g/mol
Boiling Pt: 153…157 °C (1013 hPa)
Density: 1,2281 g/cm³ (25 °C)
MDL Number: MFCD00130083
CAS Number: 14345-97-2
EINECS: 238-296-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR