2-Acetyl-1-methyl-1H-pyrrole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22044G100 AS22044G25
ACTIAS22044G100EA 0 GBP
ACTIAS22044G100 ACTIAS22044G25
2-Acetyl-1-methyl-1H-pyrrole
2-Acetyl-1-methyl-1H-pyrrole
Formula: C₇H₉NO
Boiling Pt: 200 °C (1013 hPa)
Density: 1,0445 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003089
CAS Number: 932-16-1
EINECS: 213-247-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR