2-Aminonicotinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20032G100 AS20032G25
ACTIAS20032G100EA 0 GBP
ACTIAS20032G100 ACTIAS20032G25
2-Aminonicotinic acid
2-Aminonicotinic acid
Formula: C₆H₆N₂O₂
Melting Pt: 297 °C
Density: 1.545 g/cm³ (20 °C)
MDL Number: MFCD00006318
CAS Number: 5345-47-1
EINECS: 226-296-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR