2-Amino-5-bromonicotinic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20590G100
AS20590G25
AS20590G5
ACTIAS20590G100EA
0
GBP
ACTIAS20590G100
ACTIAS20590G25
ACTIAS20590G5
2-Amino-5-bromonicotinic acid
2-Amino-5-bromonicotinic acid
Formula:
C₆H₅BrN₂O₂ MW: 217.02 g/mol |
MDL Number:
MFCD01860227 CAS Number: 52833-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...