5-Bromo-2-chloronicotinic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20413G100
AS20413G25
ACTIAS20413G100EA
0
GBP
ACTIAS20413G100
ACTIAS20413G25
5-Bromo-2-chloronicotinic acid
5-Bromo-2-chloronicotinic acid
Formula:
C₆H₃BrClNO₂ |
MDL Number:
MFCD03844847 CAS Number: 29241-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...