Potassium tetrachloroplatinate(II)

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Dipotassium tetrachloroplatinate, Potassium tetrachloroplatinate

AS23553G5 AS23553G1
ACTIAS23553G5EA 0 GBP
ACTIAS23553G5 ACTIAS23553G1
Potassium tetrachloroplatinate(II)
Potassium tetrachloroplatinate (II)
Formula: Cl₄K₂Pt
MW: 415.09 g/mol
Boiling Pt: 35…40 °C (20 mmHg)
Melting Pt: 265
Density: 3.38 g/cm³ (20 °C)
MDL Number: MFCD00011378
CAS Number: 10025-99-7
EINECS: 233-050-9
UN: 3288
ADR: 6.1 III
Merck Index: 12,07855

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR