2-Bromo-9-fluorenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-9H-fluoren-9-one

AS21480G100 AS21480G25 AS21480G5
ACTIAS21480G100EA 0 GBP
ACTIAS21480G100 ACTIAS21480G25 ACTIAS21480G5
2-Bromo-9-fluorenone
2-Bromo-9-fluorenone
Formula: C₁₃H₇BrO
MW: 259.1 g/mol
MDL Number: MFCD00010789
CAS Number: 3096-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR