4,4-Dimethyl-2-cyclohexen-1-one

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,4-Dimethylcyclohex-2-enone

AS21468G25 AS21468G5
ACTIAS21468G25EA 0 GBP
ACTIAS21468G25 ACTIAS21468G5
4,4-Dimethyl-2-cyclohexen-1-one
4,4-Dimethyl-2-cyclohexen-1-one
Formula: C₈H₁₂O
MW: 124.18 g/mol
Boiling Pt: 27.5 °C (0.2 mmHg)
Density: 0.93…0.935 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009695
CAS Number: 1073-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR