3,4-Dimethylbenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Hydroxymethyl)-o-xylene, (3,4-Dimethylphenyl)methanol

AS21986G25 AS21986G5
ACTIAS21986G25EA 125 GBP
ACTIAS21986G25 ACTIAS21986G5
3,4-Dimethylbenzyl alcohol
3,4-Dimethylbenzyl alcohol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 218
Melting Pt: 63
MDL Number: MFCD00004647
CAS Number: 6966-10-5
EINECS: 230-175-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR