3,4-Dimethylbenzylamine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20504G25 AS20504G5
ACTIAS20504G25EA 0 GBP
ACTIAS20504G25 ACTIAS20504G5
3,4-Dimethylbenzylamine
3,4-Dimethylbenzylamine
Formula: C₉H₁₃N
MDL Number: MFCD00025577
CAS Number: 102-48-7
EINECS: 203-034-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR