2,3-Dimethylbenzylamine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20506G25 AS20506G5
ACTIAS20506G25EA 0 GBP
ACTIAS20506G25 ACTIAS20506G5
2,3-Dimethylbenzylamine
2,3-Dimethylbenzylamine
Formula: C₉H₁₃N
MDL Number: MFCD00052680
CAS Number: 51586-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR