3-Hydroxybenzyl alcohol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22371G100
AS22371G25
ACTIAS22371G100EA
234
GBP
ACTIAS22371G100
ACTIAS22371G25
3-Hydroxybenzyl alcohol
3-Hydroxybenzyl alcohol
Formula:
C₇H₈O₂ Boiling Pt: 86…87 °C (0.001 mmHg) Melting Pt: 70 °C Density: 1.161 g/cm³ (25 °C) |
MDL Number:
MFCD00004643 CAS Number: 620-24-6 EINECS: 210-633-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...