3-Hydroxy-4-methoxybenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Hydroxy-4-(hydroxymethyl)anisole, 5-(Hydroxymethyl)-2-methoxyphenol, (3-Hydroxy-4-methoxyphenyl)methanol

AS22370G100 AS22370G25
ACTIAS22370G100EA 0 GBP
ACTIAS22370G100 ACTIAS22370G25
3-Hydroxy-4-methoxybenzyl alcohol
3-Hydroxy-4-methoxybenzyl alcohol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
MDL Number: MFCD00004644
CAS Number: 4383-06-6
EINECS: 224-489-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR