3-Aminobenzyl alcohol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-(Hydroxymethyl)aniline, 3-(Hydroxymethyl)aniline, 3-Aminobenzyl Alcohol, (3-Aminophenyl)methanol
AS24459G100
AS24459G25
ACTIAS24459G100EA
0
GBP
ACTIAS24459G100
ACTIAS24459G25
3-Aminobenzyl alcohol
3-Aminobenzyl alcohol
Formula:
C₇H₉NO Boiling Pt: 272 °C (1013 hPa) Melting Pt: 90…93 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007817 CAS Number: 1877-77-6 EINECS: 217-514-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...