4-(Trifluoromethoxy)benzyl alcohol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS23514G100
AS23514G25
ACTIAS23514G100EA
0
GBP
ACTIAS23514G100
ACTIAS23514G25
4-(Trifluoromethoxy)benzyl alcohol
4-(Trifluoromethoxy)benzyl alcohol
Formula:
C₈H₇F₃O₂ Boiling Pt: 53…54 °C (0,5 mmHg) Density: 1,32 g/cm³ (20 °C) |
MDL Number:
MFCD00036029 CAS Number: 1736-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...