α,α,α,5-Tetrafluoro-m-toluic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Fluoro-5-trifluoromethylbenzoic acid, 3-Fluoro-5-(trifluoromethyl)benzoic acid

AS24893G100 AS24893G25
ACTIAS24893G100EA 0 GBP
ACTIAS24893G100 ACTIAS24893G25
α,α,α,5-Tetrafluoro-m-toluic acid
α,α,α,5-Tetrafluoro-m-toluic acid
Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
MDL Number: MFCD00061293
CAS Number: 161622-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR