(3,4-Difluorophenyl)acetonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-Difluorobenzyl cyanide, 2-(3,4-Difluorophenyl)acetonitrile

AS24907G100 AS24907G25 AS24907G5
ACTIAS24907G100EA 290 GBP
ACTIAS24907G100 ACTIAS24907G25 ACTIAS24907G5
(3,4-Difluorophenyl)acetonitrile
(3,4-Difluorophenyl)acetonitrile
Formula: C₈H₅F₂N
MW: 153.13 g/mol
Boiling Pt: 110…120 °C (13 mmHg)
Density: 1.25 g/cm³ (20 °C)
MDL Number: MFCD00009974
CAS Number: 658-99-1
EINECS: 211-528-8
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR