4-Methoxy-m-toluic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20537G25
AS20537G5
ACTIAS20537G25EA
179
GBP
ACTIAS20537G25
ACTIAS20537G5
4-Methoxy-m-toluic acid
4-Methoxy-m-toluic acid
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol |
MDL Number:
MFCD00270114 CAS Number: 6880-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...