4-Methyl-m-anisic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Methoxy-4-methylbenzoic acid
AS22001G100
AS22001G25
ACTIAS22001G100EA
78
GBP
ACTIAS22001G100
ACTIAS22001G25
4-Methyl-m-anisic acid
4-Methyl-m-anisic acid
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol |
MDL Number:
MFCD00002566 CAS Number: 7151-68-0 EINECS: 230-486-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...