2-Bromo-4-(trifluoromethyl)benzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-4-(trifluoromethyl)benzoic acid, 2-Bromo-4-(trifluoromethl)benzoic acid

AS30785G1 AS30785G5
ACTIAS30785G1EA 0 GBP
ACTIAS30785G1 ACTIAS30785G5
2-Bromo-4-(trifluoromethyl)benzoic acid
2-Bromo-4-(trifluoromethyl)benzoic acid
Formula: C₈H₄BrF₃O₂
MW: 269.02 g/mol
MDL Number: MFCD07368837
CAS Number: 328-89-2
EINECS: 206-339-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR