(3-Bromophenyl)acetonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20149G100 AS20149G25
ACTIAS20149G100EA 0 GBP
ACTIAS20149G100 ACTIAS20149G25
(3-Bromophenyl)acetonitrile
(3-Bromophenyl)acetonitrile
Formula: C₈H₆BrN
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 27…28 °C
MDL Number: MFCD00001906
CAS Number: 31938-07-5
EINECS: 250-867-6
UN: 3449
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR