ω-Bromo-3-chloroacetophenone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Bromo-3'-chloroacetophenone, Bromomethyl m-chlorophenyl ketone, 2-Bromo-1-(3-chlorophenyl)ethanone, 3-Chlorophenacyl bromide, α-Bromo-3-chloroacetophenone
AS21227G25
AS21227G5
ACTIAS21227G25EA
0
GBP
ACTIAS21227G25
ACTIAS21227G5
ω-Bromo-3-chloroacetophenone
ω-Bromo-3-chloroacetophenone
Formula:
C₈H₆BrClO MW: 233.49 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00792669 CAS Number: 41011-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...