3-Ethynylaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Amino-3-ethynylbenzene, (3-Aminophenyl)acetylene, (3-Aminophenyl)ethyne, 1-Ethynyl-3-aminobenzene

AS20231G100 AS20231G25
ACTIAS20231G100EA 0 GBP
ACTIAS20231G100 ACTIAS20231G25
3-Ethynylaniline
3-Ethynylaniline
Formula: C₈H₇N
Boiling Pt: 50 °C (0.1 mmHg)
Melting Pt: 27 °C
Density: 1,04 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00014779
CAS Number: 54060-30-9
EINECS: 258-944-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR