1-Fluoro-2-ethynylbenzene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Fluorophenyl)ethyne, 1-Ethynyl-2-fluorobenzene, (2-Fluorophenyl)acetylene

AS25348G1 AS25348G25 AS25348G5
ACTIAS25348G1EA 0 GBP
ACTIAS25348G1 ACTIAS25348G25 ACTIAS25348G5
1-Fluoro-2-ethynylbenzene
1-Fluoro-2-ethynylbenzene
Formula: C₈H₅F
MW: 120.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00799540
CAS Number: 766-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR