Methyl-4-iodobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Iodobenzoic acid methyl ester, Methyl 4-iodobenzoate

AS22321G100 AS22321G25
ACTIAS22321G100EA 223 GBP
ACTIAS22321G100 ACTIAS22321G25
Methyl-4-iodobenzoate
Methyl 4-iodobenzoate
Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 112 °C
Density: 2,02 g/cm³ (10 °C)
MDL Number: MFCD00016353
CAS Number: 619-44-3
EINECS: 210-597-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR