p,α,α,α-Tetrafluoroanisole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS25342G100
AS25342G25
ACTIAS25342G100EA
0
GBP
ACTIAS25342G100
ACTIAS25342G25
p,α,α,α-Tetrafluoroanisole
p,α,α,α-Tetrafluoroanisole
Formula:
C₇H₄F₄O Boiling Pt: 104…105 °C (1013 hPa) Density: 1.323 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00040835 CAS Number: 352-67-0 EINECS: 206-523-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...