1-Iodo-3-(trifluoromethoxy)benzene
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24708G25
AS24708G5
ACTIAS24708G25EA
76
GBP
ACTIAS24708G25
ACTIAS24708G5
1-Iodo-3-(trifluoromethoxy)benzene
1-Iodo-3-(trifluoromethoxy)benzene
Formula:
C₇H₄F₃IO MW: 288.01 g/mol |
MDL Number:
MFCD01090992 CAS Number: 198206-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...