3-Fluoro-4-methoxybenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24901G100 AS24901G25
ACTIAS24901G100EA 0 GBP
ACTIAS24901G100 ACTIAS24901G25
3-Fluoro-4-methoxybenzoic acid
3-Fluoro-4-methoxybenzoic acid
Formula: C₈H₇FO₃
Melting Pt: 211.5 °C
MDL Number: MFCD00060675
CAS Number: 403-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR