4-Amino-2-fluorobenzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS23061G25
AS23061G5
ACTIAS23061G25EA
0
GBP
ACTIAS23061G25
ACTIAS23061G5
4-Amino-2-fluorobenzoic acid
4-Amino-2-fluorobenzoic acid
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol |
MDL Number:
MFCD01569397 CAS Number: 446-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...