2-Cyanobenzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22057G25
AS22057G5
ACTIAS22057G25EA
0
GBP
ACTIAS22057G25
ACTIAS22057G5
2-Cyanobenzoic acid
2-Cyanobenzoic acid
Formula:
C₈H₅NO₂ |
MDL Number:
MFCD00045796 CAS Number: 3839-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...