Methyl-2-cyanobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-cyanobenzoate, 2-Cyanobenzoic acid methyl ester

AS22058G25 AS22058G5
ACTIAS22058G25EA 235 GBP
ACTIAS22058G25 ACTIAS22058G5
Methyl-2-cyanobenzoate
Methyl 2-cyanobenzoate
Formula: C₉H₇NO₂
MW: 161.16 g/mol
MDL Number: MFCD00016376
CAS Number: 6587-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR