4-Chloro-2-methoxybenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21911G100 AS21911G25
ACTIAS21911G100EA 0 GBP
ACTIAS21911G100 ACTIAS21911G25
4-Chloro-2-methoxybenzoic acid
4-Chloro-2-methoxybenzoic acid
Formula: C₈H₇ClO₃
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 148 °C
MDL Number: MFCD00002532
CAS Number: 57479-70-6
EINECS: 260-761-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR