3-Chloro-4-methoxybenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Chloro-p-anisic acid

AS25376G1 AS25376G25 AS25376G5
ACTIAS25376G1EA 0 GBP
ACTIAS25376G1 ACTIAS25376G25 ACTIAS25376G5
3-Chloro-4-methoxybenzoic acid
3-Chloro-4-methoxybenzoic acid
Formula: C₈H₇ClO₃
MDL Number: MFCD00016512
CAS Number: 37908-96-6
EINECS: 253-708-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR