2,2-Dimethylsuccinic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,2-Dimethyl-1,4-butanedioic acid, α,α-Dimethylsuccinic acid
AS21412G100
AS21412G25
ACTIAS21412G100EA
0
GBP
ACTIAS21412G100
ACTIAS21412G25
2,2-Dimethylsuccinic acid
2,2-Dimethylsuccinic acid
Formula:
C₆H₁₀O₄ Boiling Pt: 296 °C (1013 hPa) Melting Pt: 143 °C Density: 1.323 g/cm³ (20 °C) |
MDL Number:
MFCD00004196 CAS Number: 597-43-3 EINECS: 209-899-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...