Ethyl bromopyruvate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Bromopyruvic acid ethyl ester, Ethyl 3-bromo-2-oxopropanoate, 3-Bromo-2-oxopropionic acid ethyl ester

AS51214G100 AS51214G25
ACTIAS51214G100EA 0 GBP
ACTIAS51214G100 ACTIAS51214G25
Ethyl bromopyruvate
Ethyl bromopyruvate
Formula: BrH₂CC(O)COOCH₂CH₃
MW: 195.01 g/mol
Boiling Pt: 98…100 °C (13 hPa)
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000204
CAS Number: 70-23-5
EINECS: 200-729-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR