D-(-)-Valine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-D-Val-OH, (R)-(-)-Valine

AS20411G100 AS20411G25
ACTIAS20411G100EA 0 GBP
ACTIAS20411G100 ACTIAS20411G25
D-(-)-Valine
D(-)-Valine
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 217
Melting Pt: 86
MDL Number: MFCD00064219
CAS Number: 640-68-6
EINECS: 211-368-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR