(S)-(+)-2-Amino-1-propanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS20399G100 AS20399G25
ACTIAS20399G100EA 0 GBP
ACTIAS20399G100 ACTIAS20399G25
(S)-(+)-2-Amino-1-propanol
(S)-(+)-2-Amino-1-propanol
Formula: C₃H₉NO
Boiling Pt: 173 °C (1013 hPa)
Density: 0.955 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064412
CAS Number: 2749-11-3
EINECS: 220-388-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR