3,4-Dichlorocinnamic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-Dichlorocinnamic Acid

AS24538G100 AS24538G25
ACTIAS24538G100EA 0 GBP
ACTIAS24538G100 ACTIAS24538G25
3,4-Dichlorocinnamic acid
3,4-Dichlorocinnamic acid
Formula: C₉H₆Cl₂O₂
MDL Number: MFCD00004385
CAS Number: 1202-39-7
EINECS: 214-866-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR