3,4,5-Trimethoxycinnamic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22682G100 AS22682G25
ACTIAS22682G100EA 0 GBP
ACTIAS22682G100 ACTIAS22682G25
3,4,5-Trimethoxycinnamic acid
3,4,5-Trimethoxycinnamic acid
Formula: C₁₂H₁₄O₅
Boiling Pt: 367 °C (1013 hPa)
Melting Pt: 127 °C
MDL Number: MFCD00004388
CAS Number: 90-50-6
EINECS: 201-999-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR