Tetrachloropyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,4,5,6-Tetrachloropyrimidine

AS21926G100 AS21926G25
ACTIAS21926G100EA 0 GBP
ACTIAS21926G100 ACTIAS21926G25
Tetrachloropyrimidine
Tetrachloropyrimidine
Formula: C₄Cl₄N₂
MW: 217.87 g/mol
Boiling Pt: 117…118 °C (15 mmHg)
Melting Pt: 70
MDL Number: MFCD00006062
CAS Number: 1780-40-1
EINECS: 217-228-3
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR