7-Nitroindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21148G25 AS21148G5
ACTIAS21148G25EA 95 GBP
ACTIAS21148G25 ACTIAS21148G5
7-Nitroindole
7-Nitroindole
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD00005683
CAS Number: 6960-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR