7-Methoxyindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS50147G1 AS50147G5
ACTIAS50147G1EA 0 GBP
ACTIAS50147G1 ACTIAS50147G5
7-Methoxyindole
7-Methoxyindole
Formula: C₉H₉NO
Boiling Pt: 298 °C (1013 hPa)
Density: 1,126 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00047203
CAS Number: 3189-22-8
EINECS: 221-690-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR